(4-Fluorophenyl)acetic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-(4-Fluorophenyl)acetic acid
M-Clarity™ quality level = MQ100
F13304-100G
F13304-25G
SIALF13304-100GEA
94.5
GBP
SIALF13304-100G
SIALF13304-25G
(4-Fluorophenyl)acetic acid, Sigma-Aldrich®
(4-Fluorophenyl)acetic acid
Formula:
(C₆H₄F)CH₂COOH MW: 154.14 g/mol Boiling Pt: 264 °C (1013 hPa) Melting Pt: 84…85 °C |
MDL Number:
MFCD00004343 CAS Number: 405-50-5 EINECS: 206-972-4 Merck Index: 13,04204 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...