Homovanillic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-(4-Hydroxy-3-methoxyphenyl)acetic acid, (4-Hydroxy-3-methoxyphenyl)acetic acid

002580-1G 002580-5G
FLUO002580-1GEA 0 GBP
FLUO002580-1G FLUO002580-5G
Homovanillic acid
Homovanillic acid
Formula: (C₆H₃(OH)(OCH₃))CH₂COOH
MW: 182.18 g/mol
Boiling Pt: 339 °C (1013 hPa)
Melting Pt: 145 °C
MDL Number: MFCD00004350
CAS Number: 306-08-1
EINECS: 206-176-7
Merck Index: 13,04761

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR