(5-Chloro-2-fluorophenyl)acetic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(5-Chloro-2-fluorophenyl)acetic acid

005667-1G 005667-5G
FLUO005667-1GEA 0 GBP
FLUO005667-1G FLUO005667-5G
(5-Chloro-2-fluorophenyl)acetic acid
(5-Chloro-2-fluorophenyl)acetic acid
Formula: (C₆H₃ClF)CH₂COOH
MW: 188.59 g/mol
MDL Number: MFCD01631439
CAS Number: 261762-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR