4-(Indol-3-yl)butyric acid

Supplier: EHRENSTORFER GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Indole-3-butyric acid3-Indolebutyric acid

EHERC14310000EA 142 GBP
EHERC14310000
4-(Indol-3-yl)butyric acid
4-(Indol-3-yl)butyric acid
Formula: C₁₂H₁₃NO₂
MW: 203.24 g/mol
Melting Pt: 121…126 °C
Density: 1.27 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005664
CAS Number: 133-32-4
EINECS: 205-101-5
UN: 2811
ADR: 6.1,III
Merck Index: 13,04987

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR