3-(Indol-3-yl)propionic acid
Supplier: EHRENSTORFER GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-Indolepropionic acid
, Indole-3-propionic acid
EHERC14320000EA
151
GBP
EHERC14320000
3-(Indol-3-yl)propionic acid
3-(Indol-3-yl)propionic acid
Formula:
C₁₁H₁₁NO₂ MW: 189.21 g/mol Melting Pt: 129…132 °C Density: 1.356 g/cm³ (20 °C) |
MDL Number:
MFCD00005660 CAS Number: 830-96-6 EINECS: 212-600-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...