3,4-Dimethylhexane, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger


M-Clarity™ quality level = MQ100

SIAL118419-1GEA 64.3 GBP
SIAL118419-1G
3,4-Dimethylhexane, Sigma-Aldrich®
3,4-Dimethylhexane
Formula: C₈H₁₈
MW: 114.23 g/mol
Boiling Pt: 118 °C (1013 hPa)
Melting Pt: –64 °C
Density: 0.7152 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00039927
CAS Number: 583-48-2
EINECS: 209-504-7
UN: 1262
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR