2,6,10,14-Tetramethylpentadecane, SAFC®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Pristane

M-Clarity™ quality level = MQ200

P9622-10X1ML P9622-5X5ML
SIALP9622-10X1MLEA 229 GBP
SIALP9622-10X1ML SIALP9622-5X5ML
2,6,10,14-Tetramethylpentadecane, SAFC®
2,6,10,14-Tetramethylpentadecane
Formula: C₁₉H₄₀
MW: 268.53 g/mol
Boiling Pt: 296 °C (1013 hPa)
Density: 0.7845 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008952
CAS Number: 1921-70-6
EINECS: 217-650-8
Merck Index: 12,07932

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR