2,6,10,14-Tetramethylpentadecane, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Pristane
M-Clarity™ quality level = MQ200
P2870-100ML
P2870-250ML
P2870-500ML
SIALP2870-100MLEA
986
GBP
SIALP2870-100ML
SIALP2870-250ML
SIALP2870-500ML
2,6,10,14-Tetramethylpentadecane, Sigma-Aldrich®
2,6,10,14-Tetramethylpentadecane
Formula:
C₁₉H₄₀ MW: 268.53 g/mol Boiling Pt: 296 °C (1013 hPa) Density: 0.7845 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008952 CAS Number: 1921-70-6 EINECS: 217-650-8 Merck Index: 12,07932 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...