1,1,3,3-Tetramethylbutyl isothiocyanate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
009556-10G
009556-50G
009556-25G
009556-5G
FLUO009556-10GEA
0
GBP
FLUO009556-10G
FLUO009556-50G
FLUO009556-25G
FLUO009556-5G
1,1,3,3-Tetramethylbutyl isothiocyanate
1,1,3,3-Tetramethylbutyl isothiocyanate
Formula:
C₉H₁₇NS MW: 171.31 g/mol |
MDL Number:
MFCD00004817 CAS Number: 17701-76-7 EINECS: 241-710-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...