Ethyl pivalate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Trimethylacetic acid ethyl ester, Ethyltrimethylacetate, Pivalic acid ethyl ester, 2,2-Dimethylpropionic acid ethyl ester, Ethyl trimethylacetate

003884-100G 003884-500G
FLUO003884-100GEA 0 GBP
FLUO003884-100G FLUO003884-500G
Ethyl pivalate
Ethyl pivalate
Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 118 °C (1013 hPa)
Melting Pt: –90 °C
Density: 0.86 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00009129
CAS Number: 3938-95-2
EINECS: 223-520-1
UN: 3272
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR