L(-)-Glutathione (reduced form)

Supplier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: L-Glutathione ReducedGlutathione Reduced(S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-mercapto-1-oxopropan-2-yl)amino)-5-oxopentanoic acidGSH

SERA23150.04EA 223.5 GBP
SERA23150.04 SERA23150.02 SERA23150.03
L(-)-Glutathione (reduced form)
L(-)-Glutathione (reduced form)
Formula: C₁₀H₁₇N₃O₆S
MW: 307.33 g/mol
Melting Pt: 185…195 °C
Density: 1.475 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00065939
CAS Number: 70-18-8
EINECS: 200-725-4
Merck Index: 13,04488

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR