n-Tetracosane, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
N-tetracosane
M-Clarity™ quality level = MQ100
SIALT8752-25GEA
92
GBP
SIALT8752-25G
n-Tetracosane, Sigma-Aldrich®
n-Tetracosane
Formula:
H₃C(CH₂)₂₂CH₃ MW: 338.66 g/mol Boiling Pt: 391.3 °C (1013 hPa) Melting Pt: 49…51 °C Density: 0.7787 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00009352 CAS Number: 646-31-1 EINECS: 211-474-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...