n-Octane
Supplier: EHRENSTORFER GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
EHERC15711000EA
87.3
GBP
EHERC15711000
n-Octane
n-Octane
Formula:
H₃C(CH₂)₆CH₃ MW: 114.23 g/mol Boiling Pt: 126 °C (1013 hPa) Melting Pt: –57 °C Density: 0.703 g/cm³ (20 °C) Flash Pt: 8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009556 CAS Number: 111-65-9 EINECS: 203-892-1 UN: 1262 ADR: 3,II Merck Index: 13,06782 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...