1-Octanesulphonic acid sodium salt

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Sodium octane-1-sulphonate

009637-10G 009637-25G
FLUO009637-10GEA 0 GBP
FLUO009637-10G FLUO009637-25G
1-Octanesulphonic acid sodium salt
1-Octanesulphonic acid sodium salt
Formula: C₈H₁₇NaO₃S
MW: 216.28 g/mol
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007544
CAS Number: 5324-84-5
EINECS: 226-195-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1244cb11db07775dec2e193823e40080024b7036&vid=32d95cd9e321818e2f94c8261c4b4e6180ee0fc7&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR