Methanesulphonic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
M-Clarity™ quality level = MQ200
SIAL471356-5MLEA
73.3
GBP
SIAL471356-5ML
SIAL471356-100ML
NOVS471356-100ML
SIAL471356-500ML
Methanesulphonic acid, Sigma-Aldrich®
Methanesulphonic acid
Formula:
CH₃SO₃H MW: 96.11 g/mol Boiling Pt: 217 °C (1013 hPa) Melting Pt: 20 °C Density: 1.48 g/cm³ (20 °C) Flash Pt: 189 °C (closed cup) Storage Temperature: Refrigerator |
MDL Number:
MFCD00007518 CAS Number: 75-75-2 EINECS: 200-898-6 UN: 2586 ADR: 8,III Merck Index: 13,05981 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...