Lithium dodecyl sulphate

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Dodecyl sulphuric acid lithium salt, Dodecyl sulphate lithium salt, Lithium lauryl sulphate, Lauryl sulphate lithium salt

EMR164010 EMR164050 EMR164100 EMR164250
EURLEMR164010EA 0 GBP
EURLEMR164010 EURLEMR164050 EURLEMR164100 EURLEMR164250
Lithium dodecyl sulphate
Lithium dodecyl sulphate
Formula: C₁₂H₂₅LiO₄S
MW: 272.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007467
CAS Number: 2044-56-6
EINECS: 218-058-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR