1,10-Decanediol, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1,10-Dihydroxy decane
M-Clarity™ quality level = MQ200
D1203-100G
D1203-500G
SIALD1203-100GEA
57.5
GBP
SIALD1203-100G
SIALD1203-500G
1,10-Decanediol, Sigma-Aldrich®
1,10-Decanediol
Formula:
C₁₀H₂₂O₂ MW: 174.28 g/mol Boiling Pt: 297 °C (1013 hPa) Melting Pt: 72 °C Density: 0.883 g/cm³ (80 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004749 CAS Number: 112-47-0 EINECS: 203-975-2 Merck Index: 13,02869 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...