1-Tetradecanol, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-Tetradecyl alcohol
, 1-Hydroxy tetradecane
, Myristyl alcohol
M-Clarity™ quality level = MQ200
SIAL185388-100GEA
56.2
GBP
SIAL185388-100G
1-Tetradecanol, Sigma-Aldrich®
1-Tetradecanol
Formula:
H₃C(CH₂)₁₂CH₂OH MW: 214.39 g/mol Boiling Pt: 289 °C (1013 hPa) Melting Pt: 37…39 °C Density: 0.8 g/cm³ (60 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004757 CAS Number: 112-72-1 EINECS: 204-000-3 Merck Index: 12,06418 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...