Spermidine

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: N1-(3-Aminopropyl)butane-1,4-diamineN-(3-Aminopropyl)-1,4-diaminobutane4-Azaoctamethylenediamine

MOLEM24358880EA 0 GBP
MOLEM24358880
Spermidine
Spermidine
Formula: C₇H₁₉N₃
MW: 145.25 g/mol
Boiling Pt: 246 °C (1013 hPa)
Melting Pt: 20…25 °C
Density: 0,925 g/cm³ (20 °C)
MDL Number: MFCD00008229
CAS Number: 124-20-9
EINECS: 204-689-0
Merck Index: 12,08893

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR