Dioctylamine 97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Di-n-octylamine, N-Octyl-1-octanamine
117080250
117081000
117085000
ACRO117080250EA
24.1
GBP
ACRO117080250
ACRO117081000
ACRO117085000
Dioctylamine 97%
Dioctylamine
Formula:
C₁₆H₃₅N MW: 241.46 g/mol Boiling Pt: 297…298 °C (760 mmHg) Melting Pt: 14…15 °C Density: 0.79 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009557 CAS Number: 1120-48-5 EINECS: 214-311-6 UN: 2735 ADR: 8,III |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4412 to 1.4432 (20°C, 589 nm) |
GC | ≥96.0 % |
Residue after ignition | ≤0.1 % |
Color of alcohol solution | ≤60 APHA |
Color of a HCl solution | Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...