1,5-Diaminopentane

Supplier: ABCR GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Cadaverine1,5-PentanediamineCadavarinePentamethylendiamine

ABCRAB133280.0500EA 0 GBP
ABCRAB133280.0500
1,5-Diaminopentane
1,5-Diaminopentane
Formula: C₅H₁₄N₂
MW: 102.18 g/mol
Boiling Pt: 178 °C (1013 hPa)
Density: 0.9174 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008239
CAS Number: 462-94-2
EINECS: 207-329-0
UN: 2735
ADR: 8,III
Merck Index: 12,01645

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR