1-Propyl isothiocyanate

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

P2520.10 P2520.50
XXXXP2520.10EA 0 GBP
XXXXP2520.10 XXXXP2520.50
1-Propyl isothiocyanate
1-Propyl isothiocyanate
Formula: C₄H₇NS
MW: 101.17 g/mol
Boiling Pt: 152…153 °C (1013 hPa)
Density: 0,9909 g/cm³ (0 °C)
MDL Number: MFCD00004823
CAS Number: 628-30-8
EINECS: 211-035-8
UN: 2924
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR