Silver trifluoromethanesulphonate ≥99%

Supplier: Strem Chemicals, Inc.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Trifluoromethanesulphonic acid silver saltSilver triflateSilver trifluoromethanesulphonate

STRE47-2000-5GEA 0 GBP
STRE47-2000-5G
Silver trifluoromethanesulphonate ≥99%
Silver trifluoromethanesulphonate

Metal Triflate

Formula: CAgF₃O₃S
MW: 256.94 g/mol
Melting Pt: 356 °C
MDL Number: MFCD00013226
CAS Number: 2923-28-6
EINECS: 220-882-2
UN: 3260
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR