1,1,1,2,2-Pentafluoro-4-iodobutane

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Iodo-3,3,4,4,4-pentafluorobutane

010502-25G 010502-1G 010502-5G
FLUO010502-25GEA 0 GBP
FLUO010502-25G FLUO010502-1G FLUO010502-5G
1,1,1,2,2-Pentafluoro-4-iodobutane
1,1,1,2,2-Pentafluoro-4-iodobutane
Formula: C₄H₄F₅I
MW: 273.97 g/mol
MDL Number: MFCD00155910
CAS Number: 40723-80-6
EINECS: 255-055-5
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR