EDTA (ethylenediamine tetraacetic acid), Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Kestranal® A, OmniPur® EDTA, Edetic acid, Ethylenediaminetetraacetic acid, Ethylenedinitrilotetra-acetic acid, 2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid, (Ethylenedinitrilo)tetraacetic acid, Idranal® II
M-Clarity™ quality level = MQ200
03620-1KG
03620-250G
03620-50G
SIAL03620-1KGEA
121
GBP
SIAL03620-1KG
SIAL03620-250G
SIAL03620-50G
EDTA (ethylenediamine tetraacetic acid), Sigma-Aldrich®
EDTA (Ethylenediamine tetraacetic acid)
Formula:
C₁₀H₁₆N₂O₈ MW: 292.25 g/mol Boiling Pt: 614 °C (1013 hPa) Melting Pt: 220 °C Density: 0.86 g/cm³ (20 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003541 CAS Number: 60-00-4 EINECS: 200-449-4 REACH: 01-2119486399-18 Merck Index: 13,03546 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...