Adipic acid
Supplier: EHRENSTORFER GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1,6-Hexanedioic acid
, Hexanedioc acid
EHERC10045900EA
99.1
GBP
EHERC10045900
Adipic acid
Adipic acid
Formula:
HOOC(CH₂)₄COOH MW: 146.14 g/mol Boiling Pt: 337 °C (1013 hPa) Melting Pt: 151 °C Density: 1.36 g/cm³ (25 °C) Flash Pt: 196 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00004420 CAS Number: 124-04-9 EINECS: 204-673-3 Merck Index: 13,00163 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...