Mercaptoacetic acid (thioglycolic acid)

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Mercaptoacetic acid

MOLE14299627EA 0 GBP
MOLE14299627
Mercaptoacetic acid (thioglycolic acid)
Mercaptoacetic acid (Thioglycolic acid)
Formula: HSCH₂COOH
MW: 92.12 g/mol
Boiling Pt: 96 °C (1013 hPa)
Melting Pt: –16.5 °C
Density: 1.33 g/cm³ (20 °C)
Flash Pt: 130 °C (closed cup)
Storage Temperature: Refrigerator
MDL Number: MFCD00004876
CAS Number: 68-11-1
EINECS: 200-677-4
UN: 1940
ADR: 8,II
Merck Index: 12,09472
https://uk.vwr.com/stibo/web/std.lang.all/90/10/2299010.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR