Glycine, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Aminoacetic acidH-Gly-OHGlycocoll

M-Clarity™ quality level = MQ100

94119-10GEA 45.1 GBP
94119-10G
Glycine, Supelco®
Glycine
Formula: NH₂CH₂COOH
MW: 75.07 g/mol
Boiling Pt: 181 °C (1013 hPa)
Melting Pt: 83 °C
Density: 1.64 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008131
CAS Number: 56-40-6
EINECS: 200-272-2
Merck Index: 13,04504
https://uk.vwr.com/stibo/web/std.lang.all/98/49/27079849.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR