Magnesium stearate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Stearic acid Magnesium(II) saltStearic acid magnesium saltMagnesium(II) Stearate

SIAL26454-1KGEA 93.3 GBP
SIAL26454-1KG
Magnesium stearate, Sigma-Aldrich®
Magnesium stearate
Formula: C₃₆H₇₀MgO₄
MW: 591.25 g/mol
Melting Pt: 130…140 °C
Density: 1.03 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036391
CAS Number: 557-04-0
EINECS: 209-150-3
Merck Index: 13,05714

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR