Lithium stearate
Supplier: Roth Carl
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Stearic acid lithium salt
, Lithium Stearate
ROTH8238.1EA
27.7
GBP
ROTH8238.1
Lithium stearate
Lithium stearate
Formula:
C₁₈H₃₅LiO₂ MW: 290.42 g/mol Melting Pt: 220…221.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042032 CAS Number: 4485-12-5 EINECS: 224-772-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...