Butyric acid, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Butanoic acid

M-Clarity™ quality level = MQ100

19215-5MLEA 95.1 GBP
19215-5ML
Butyric acid, Supelco®
Butyric acid
Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 163.5 °C (1013 hPa)
Melting Pt: –3 °C
Density: 0.958 g/cm³ (20 °C)
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00002814
CAS Number: 107-92-6
EINECS: 203-532-3
UN: 2820
ADR: 8,III
Merck Index: 13,01593

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR