Diethyl malonate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1,3-Propanedioic acid diethyl ester, Malonic acid diethyl ester

M-Clarity™ quality level = MQ300

W237507-10KG W237507-1KG W237507-25KG W237507-SAMPLE
SIALW237507-10KGEA 332 GBP
SIALW237507-10KG SIALW237507-1KG SIALW237507-25KG SIALW237507-SAMPLE
Diethyl malonate, Sigma-Aldrich®
Diethyl malonate
Formula: C₇H₁₂O₄
MW: 160.17 g/mol
Boiling Pt: 199 °C (1013 hPa)
Melting Pt: –50 °C
Density: 1.055 g/cm³ (20 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00009195
CAS Number: 105-53-3
EINECS: 203-305-9
Merck Index: 13,03856

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR