Dibutyl sebacate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Sebacic acid dibutyl ester, 1,10-Decanedioic acid dibutyl ester, Dibutyl decanedioate
84840-1L
84840-250ML
SIAL84840-1LEA
159
GBP
SIAL84840-1L
SIAL84840-250ML
Dibutyl sebacate, Sigma-Aldrich®
Dibutyl sebacate
Formula:
C₁₈H₃₄O₄ MW: 314.47 g/mol Boiling Pt: 180 °C (1013 hPa) Melting Pt: –11 °C Density: 0.94 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00027218 CAS Number: 109-43-3 EINECS: 203-672-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...