Methyl valerate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Valeric acid methyl ester, Methyl Pentanoate

M-Clarity™ quality level = MQ100

W275204-4KG-K W275204-9KG-K
SIALW275204-4KG-KEA 478 GBP
SIALW275204-4KG-K SIALW275204-9KG-K
Methyl valerate, Sigma-Aldrich®
Methyl valerate
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 126.5 °C (1013 hPa)
Melting Pt: –91 °C
Density: 0.89 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009478
CAS Number: 624-24-8
EINECS: 210-838-0
UN: 3272
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR