Acetanilide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: N-Phenylacetamide

M-Clarity™ quality level = MQ100

SIAL00401-5GEA 77.9 GBP
SIAL00401-5G
Acetanilide, Sigma-Aldrich®
Acetanilide
Formula: C₈H₉NO
MW: 135.17 g/mol
Boiling Pt: 304 °C (1013 hPa)
Melting Pt: 115 °C
Density: 1.22 g/cm³ (20 °C)
Flash Pt: 174 °C
MDL Number: MFCD00008674
CAS Number: 103-84-4
EINECS: 203-150-7
Merck Index: 13,00051

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR