N-(2-Acetamido)-2-aminoethanesulphonic acid (ACES), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: N-(Carbamoylmethyl)-2-aminoethanesulphonic AcidN-(Carbamoylmethyl)taurineACES

M-Clarity™ quality level = MQ100

SIAL00194-100GEA 519 GBP
SIAL00194-100G
N-(2-Acetamido)-2-aminoethanesulphonic acid (ACES), Sigma-Aldrich®
N-(2-Acetamido)-2-aminoethanesulphonic acid (ACES)
Formula: H₂NCOCH₂NHCH₂CH₂SO₃H
MW: 182.2 g/mol
Melting Pt: 270…295 °C
Storage Temperature: Ambient
MDL Number: MFCD00008030
CAS Number: 7365-82-4
EINECS: 230-908-4
Merck Index: 13,00037
https://uk.vwr.com/stibo/web/std.lang.all/40/46/2354046.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR