Formamide, Spectral Grade

Supplier: IBI SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Carbamaldehyde

SHETIB72020EA 0 GBP
SHETIB72020 SHETIB72024
Formamide, Spectral Grade
Formamide

Spectral Grade Formamide is used as a denaturant in hybridization and electrophoresis applications. Formamide is used to denature DNA and RNA. It separates the two strands of the double helix.
Formamide is supplied distilled, deionized and packaged under nitrogen


The purity of IBI formamide is determined by way of gas chromatography. Store at +4C in the dark and warm to room temperature prior to opening.


When exposed to oxygen, formamide breaks down into formic acid and ammonia, which can degrade nucleic acid samples. Bottles that have been opened and stored should be deionized prior to use by mixing 1gm of mixed bed resin per 10ml of formamide for 30 minutes.

Formula: HCONH₂
MW: 45.04 g/mol
Boiling Pt: 210 °C (1013 hPa)
Melting Pt: 2 °C
Density: 1.1334 g/cm³ (20 °C)
Flash Pt: 79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007941
CAS Number: 75-12-7
EINECS: 200-842-0
Merck Index: 13,04261
https://uk.vwr.com/stibo/web/std.lang.all/25/02/2302502.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR