2-Oxoglutaric acid

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 2-Ketoglutaric acid2-Oxopentanedioic acidA-Ketoglutaric acidα-Ketoglutaric acid

MOLE12974102-100GEA 0 GBP
MOLE12974102-100G
2-Oxoglutaric acid
2-Oxoglutaric acid
Formula: HOOCCH₂CH₂COCOOH
MW: 146.1 g/mol
Boiling Pt: 323 °C (1013 hPa)
Melting Pt: 112…116 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004165
CAS Number: 328-50-7
EINECS: 206-330-3
Merck Index: 13,05320

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR