2-Oxoglutaric acid
Supplier: PanReac AppliChem
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
2-Ketoglutaric acid
, 2-Oxopentanedioic acid
, A-Ketoglutaric acid
, α-Ketoglutaric acid
A1138.0250EA
629
GBP
A1138.0250
2-Oxoglutaric acid
2-Oxoglutaric acid
Formula:
HOOCCH₂CH₂COCOOH MW: 146.1 g/mol Boiling Pt: 323 °C (1013 hPa) Melting Pt: 112…116 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004165 CAS Number: 328-50-7 EINECS: 206-330-3 Merck Index: 13,05320 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...