Di-tert-butyl pyrocarbonate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Boc Anhydride, Di-tert-butyl dicarbonate, DIBOC

021896-1KG 021896-500G
FLUO021896-1KGEA 0 GBP
FLUO021896-1KG FLUO021896-500G
Di-tert-butyl pyrocarbonate
Di-tert-butyl pyrocarbonate
Formula: C₁₀H₁₈O₅
MW: 218.25 g/mol
Boiling Pt: 235 °C (1013 hPa)
Melting Pt: 20…23 °C
Density: 1.02 g/cm³ (20 °C)
Flash Pt: 37 °C
Storage Temperature: Ambient
MDL Number: MFCD00008805
CAS Number: 24424-99-5
EINECS: 246-240-1
UN: 2930
ADR: 6.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae68e9d9dc1f713c2e5ac96bd05040cec5c7bcdd&vid=719e05eb5ef4a6a4176af5e827ff4390d03d64ab&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR