Methyl ethyl ketone analytical standard, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 2-ButanoneMEKEK 500 AdditiveButanoneEthyl methyl ketone

442339.EA 179 GBP
442339.
Methyl ethyl ketone analytical standard, Supelco®
Methyl ethyl ketone
Formula: C₂H₅COCH₃
MW: 72.11 g/mol
Boiling Pt: 79.6 °C (1013 hPa)
Melting Pt: –86 °C
Density: 0.8054 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Freezer
MDL Number: MFCD00011648
CAS Number: 78-93-3
EINECS: 201-159-0
UN: 1193
ADR: 3,II
REACH: 01-2119457290-43
Merck Index: 13,06097
https://uk.vwr.com/stibo/web/std.lang.all/03/79/720379.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR