2-Pentadecanone, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Pentadecan-2-one

M-Clarity™ quality level = MQ300

W372404-100G-K W372404-1KG-K
SIALW372404-100G-KEA 93.9 GBP
SIALW372404-100G-K SIALW372404-1KG-K
2-Pentadecanone, Sigma-Aldrich®
2-Pentadecanone
Formula: C₁₅H₃₀O
MW: 226.4 g/mol
Density: 0.82 g/cm³ (39 °C)
Storage Temperature: Ambient
MDL Number: MFCD00053712
CAS Number: 2345-28-0
EINECS: 219-064-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR