2'-Chloro-4'-fluoroacetanilide
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
N-(2-Chloro-4-fluorophenyl)acetamide
FL3155.5
FL3155.25
FL3155.100
CHMPFL3155.5EA
0
GBP
CHMPFL3155.5
CHMPFL3155.25
CHMPFL3155.100
2'-Chloro-4'-fluoroacetanilide
2'-Chloro-4'-fluoroacetanilide
Formula:
C₈H₇ClFNO MW: 187.6 g/mol Boiling Pt: 332 °C (1013 hPa) Melting Pt: 117 °C |
MDL Number:
MFCD00042594 CAS Number: 399-35-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...