1-Chloro-3-pentanone, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ200

C62506-25G
SIALC62506-25GEA 395 GBP
SIALC62506-25G
1-Chloro-3-pentanone, Sigma-Aldrich®
1-Chloro-3-pentanone
Formula: C₅H₉ClO
MW: 120.58 g/mol
Storage Temperature: Refrigerator
CAS Number: 32830-97-0
EINECS: 251-247-8
UN: 1224
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR