Ethyl bromofluoroacetate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Bromofluoroacetic ethyl ester, Ethylbromofluoroacetate

003215-10G 003215-50G 003215-100G
FLUO003215-10GEA 0 GBP
FLUO003215-10G FLUO003215-50G FLUO003215-100G
Ethyl bromofluoroacetate
Ethyl bromofluoroacetate
Formula: BrFCHCOOCH₂CH₃
MW: 184.99 g/mol
Boiling Pt: 154 °C (1013 hPa)
Density: 1.55866 g/cm³ (17 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00042095
CAS Number: 401-55-8
EINECS: 206-928-4
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR