Bromoacetic acid 97%, Reagent Grade, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
M-Clarity™ quality level = MQ200
B56307-100G
B56307-2KG
B56307-500G
SIALB56307-100GEA
68.2
GBP
SIALB56307-100G
SIALB56307-2KG
SIALB56307-500G
Bromoacetic acid 97%, Reagent Grade, Sigma-Aldrich®
Bromoacetic acid
Formula:
BrCH₂COOH MW: 138.95 g/mol Boiling Pt: 206 °C (1013 hPa) Melting Pt: 46…49 °C Density: 1.9335 g/cm³ (20 °C) Flash Pt: >112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002678 CAS Number: 79-08-3 EINECS: 201-175-8 UN: 3425 ADR: 8,II Merck Index: 13,01384 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...