(S)-(+)-4-Penten-2-ol, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(S)-(+)-1-Penten-4-ol, (S)-(+)-Allyl methyl carbinol, (S)-Pent-4-en-2-ol
M-Clarity™ quality level = MQ100
558028-1G
558028-5G
SIAL558028-1GEA
185
GBP
SIAL558028-1G
SIAL558028-5G
(S)-(+)-4-Penten-2-ol, Sigma-Aldrich®
(S)-(+)-4-Penten-2-ol
Formula:
C₅H₁₀O MW: 86.13 g/mol |
MDL Number:
MFCD03427200 CAS Number: 55563-79-6 UN: 1987 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...