D-(+)-Tryptophan
Supplier: Molekula
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
H-D-Trp-OH
MOLEM38891077EA
0
GBP
MOLEM38891077
D-(+)-Tryptophan
D(+)-Tryptophan
Formula:
C₁₁H₁₂N₂O₂ MW: 204.23 g/mol Melting Pt: 280…283 °C |
MDL Number:
MFCD00005647 CAS Number: 153-94-6 EINECS: 205-819-9 Merck Index: 14,09797 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...