Manganese(II) chloride tetrahydrate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Manganese chloride tetrahydrate
, Manganous chloride tetrahydrate
, Manganese Chloride, Tetrahydrate
, Manganese(II) chloride tetrahydrate
, Manganese dichloride tetrahydrate
, Manganese chloride (tetrahydrate)
M-Clarity™ quality level = MQ200
SIALM5005-100GEA
64
GBP
SIALM5005-100G
Manganese(II) chloride tetrahydrate, Sigma-Aldrich®
Manganese (II) chloride tetrahydrate
Formula:
MnCl₂·4H₂O MW: 197.9 g/mol Boiling Pt: 1190 °C (1013 hPa) Melting Pt: 58…59 °C Density: 2.01 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149792 CAS Number: 13446-34-9 EINECS: 231-869-6 UN: 3077 ADR: 9,III Merck Index: 13,05751 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...