Cerium(IV) sulphate tetrahydrate

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Ceric sulphate tetrahydrateCerium disulphate tetrahydrate

ROTHP014.1EA 147.58 GBP
ROTHP014.1
Cerium(IV) sulphate tetrahydrate
Cerium (IV) sulphate tetrahydrate
Formula: Ce(SO₄)₂·4H₂O
MW: 404.3 g/mol
Melting Pt: 180…200 °C
Density: 3.91 g/cm³ (20 °C)
MDL Number: MFCD00149427
CAS Number: 10294-42-5
EINECS: 237-029-5
UN: 3260
ADR: 8,III
Merck Index: 12,02036

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR